Learn More
ZeptoMetrix SARS-CoV-2 (Isolate: Italy-INMI1) Culture Fluid (Heat Inactivated)
SDP
Catalog No. 22158377 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-377 1 mL
1 options
Catalog No. 22-158-377 Supplier ZeptoMetrix Supplier No. 0810589CFHI
Add to Cart
Add to Cart
SARS-CoV-2 (Isolate: Italy-INMI1) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus.
  • Heat inactivated viral culture fluids are sold as consumable testing materials
  • The suitability and performance characteristics should be determined by your laboratory for each intended usage
  • Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays
Specifications
Content And Storage Repetitive freezing and thawing is not recommended (aliquot material if necessary). Titer will be altered by multiple freeze-thaws.
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Type Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Includes Each frozen aliquot contains 1 mL of titered viral culture fluid.
Quantity 1 mL