Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid (Heat Inactivated)
SDP

Catalog No. 22158369 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
0.5 mL
1 mL
Includes:
Each frozen aliquot contains 0.50 mL of heat inactivated viral culture fluid
Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Includes
22-158-369 1 mL Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid
22-158-364 0.5 mL Each frozen aliquot contains 0.50 mL of heat inactivated viral culture fluid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-158-369 Supplier ZeptoMetrix Supplier No. 0810587CFHI
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus.

  • Heat inactivated viral culture fluids are sold as consumable testing materials
  • The suitability and performance characteristics should be determined by your laboratory for each intended usage
  • Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays

Specifications

Content And Storage Heat inactivated viral culture fluids should be stored at -65°C or below
Description Repetitive freezing and thawing is not recommended (aliquot material if necessary)
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents
Type Culture Fluid
For Use With (Application) Isolate: USA-WA1/2020
Includes Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.