Learn More
Zeptometrix SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid (UV Inactivated)
Shop All ZeptoMetrix Corporation Products

Description
Specifications
Specifications
| Certifications/Compliance | ISO 13485 |
| Content And Storage | Store at -65°C or below |
| For Use With (Application) | Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays |
| Quantity | 1 mL |
For the purchase of Zeptometrix's BSL2 or BSL3 level products the Laboratories Biosafety Level must be consistent with the products requirement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.