Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zeptometrix SARS-CoV-2 Lineage P1;Gamma Variant (Japan/TY7-503/2021) Culture Fluid (Heat Inactivated)
SDP

Catalog No. 22158425 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-425 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-425 Supplier Zeptometrix Supplier No. 0810616CFHI
Add to Cart
Add to Cart

Infectious virus preparation designed to meet research and product development needs.

SARS-Related Coronavirus 2 Lineage P.1; Gamma Variant (Isolate: Japan/TY7-503/2021) is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus. Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2), isolate hCoV-19/Japan/TY7-503/2021 (Brazil P.1) (also referred to as TY7-503, hCoV19/Japan/IC-0564/2021) was isolated in airport quarantine in Japan from a COVID-19 positive passenger from Brazil in January 2021. Under the nomenclature system introduced by GISAID (Global Initiative on Sharing All Influenza Data), SARS-CoV-2, isolate hCoV19/Japan/TY7-503/2021 (Brazil P.1) is assigned lineage P.1 and GISAID clade GR using Phylogenetic Assignment of Named Global Outbreak LINeages (PANGOLIN) tool. The complete genome of SARS-CoV2, isolate hCoV-19/Japan/TY7-503/2021 (Brazil P.1) has been sequenced [Clinical isolate (referred to as hCoV19/Japan/IC-0564/2021): GISAID: EPI_ISL_792683; Passage 2 in Vero E6/TMPRSS2 cells: GISAID: EPI_ISL_877769]. Each frozen aliquot contains 1 mL of heat inactivated viral culture fluid. The culture fluid was clarified prior to heat inactivation by centrifugation to remove cellular debris. The pre-inactivation titer was determined from an infectious aliquot. Viral inactivation is verified after heat inactivation by the absence of viral growth in tissue culture-based infectivity assays.

Specifications

Certifications/Compliance ISO 13485
Content And Storage Store at -65°C or below
Type Culture Fluid
For Use With (Application) Nucleic Acid-Molecular Testing, Limit of Detection (LOD) Studies, Cross-reactivity Studies, Other Viral-based Assays
Quantity 1 mL

For the purchase of Zeptometrix's BSL2 or BSL3 level products the Laboratories Biosafety Level must be consistent with the products requirement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.