Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LumiraDx SARS-CoV-2 Liquid Control Kit

Catalog No. 22181002 Shop All Roche Diagnostics Products
Change view
Click to view available options
Sample Type:
Nasal Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
22-181-002 Nasal Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-181-002 Supplier LumiraDx Supplier No. 10418214001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Positive and negative quality controls to complete QC assessment of LumiraDx instrument and LumiraDx SARS-CoV-2 Ag test strips

  • Features:
    • Liquid quality controls to be used with the LumiraDx instrument and LumiraDx SARS-CoV-2 Ag test
  • Includes:
    • Two 0.5 mL vial SARS-CoV-2 Ag positive control
    • Two 0.5 mL vial SARS-CoV-2 Ag negative control
    • 24 transfer pipettes (20 μL)
    • LumiraDx SARS-CoV-2 Ag quality control pack insert
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Nasal Swab
Type External QC
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.