Learn More
AdipoGen anti-SARS-CoV-2 Spike Protein S1 mAb (rec.) (blocking) (Covi-2) (preservative free) (100 µg)
SDP
Supplier:  AdipoGen AG27B6006PFC100
Encompass_Preferred
Recombinant Antibody. Recognizes and binds to recombinant SARS-CoV-2 Spike Protein S1. It blocks the interaction of SARS-CoV-2 Spike Protein S1 (RBD) to human ACE2. Detects a different epitope as anti-SARS-CoV-2 Spike Protein S1, mAb (rec.) (blocking) (Covi-1) (Prod. No. AG-27B-6005PF https://adipogen.com/ag-27b-6005pf-anti-sars-cov-2-spike-protein-s1-mab-rec-blocking-covi-1-preservative-free.html ). Applications: ELISA, FUNC (Blocking), WB. Source: HEK 293 cells. Liquid. In PBS. The respiratory syndrome beta-coronavirus SARS-CoV-2 causes an epidemic of acute respiratory syndrome (COVID-19). SARS-CoV-2 contains 4 structural proteins, including Envelope (E), Membrane (M), Nucleocapsid (N) and Spike (S), composed of two subunits S1 and S2. The S1 subunit contains a receptor binding domain (RBD), which binds to the cell surface receptor Angiotensin-Converting Enzyme 2 (ACE2) present at the surface of epithelial cells, causing mainly infection of human respiratory cells.
Catalog No. 50-205-3052