Learn More
American Research Products Inc SARS CoV 2 N ELISA Kit
Catalog No. 5000327497
Encompass
SARS-CoV-2 N ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma swabs Method Sandwich
Catalog No. 50-003-27497 Supplier American Research Products Inc Supplier No. CSBEL33251
SARS-CoV-2 N ELISA Kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma swabs Method Sandwich