Learn More
Rockland Immunochemicals SARS-CoV-2 N Protein 100µg
SDP
Supplier:  Rockland Immunochemicals 200401MS40100
Encompass
Anti-SARS-CoV-2 Nucleocapsid (N) Protein Antibody has been tested for use in Western Blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 47 kDa in size corresponding to SARS-CoV-2 Nucleocapsid (N) protein by western blotting in the appropriate cell lysate or extract.
Catalog No. 50-202-4058