Learn More
AdipoGen SARS-CoV-2 NeutrAbs B16172
SDP
Supplier:  AdipoGen AG48B0007KI01
Encompass_Preferred
Screening Assay. To measure the presence of neutralizing/blocking antibodies in human serum/plasma that inhibit the binding of the SARS-CoV-2 Spike RBD B.1.617.2 Variant, Delta protein to its human receptor ACE2. Coronaviruses CoVs are enveloped non-segmented positive-sense single-stranded RNA viruses and can infect respiratory, gastrointestinal, hepatic and central nervous system of human and many wild animals. Recently, a new severe acute respiratory syndrome beta-coronavirus called SARS-CoV-2 has emerged, which causes an epidemic of acute respiratory syndrome called coronavirus human disease 2019 or COVID-19. SARS-CoV-2 contains 4 structural proteins, including Envelope E, Membrane M, Nucleocapsid N and Spike S, which is a transmembrane protein, composed of two subunits S1 and S2.
Catalog No. 50-256-2722