Learn More
Genscript Corporation SARS-CoV-2 (Omicron) Neutralizing Antibody Standard
Supplier:  Genscript Corporation A0216120
Encompass
The product is specific for SARS-CoV-2 RBD domain. The product can neutralize Wild-Type SARS-CoV-2 and Variants of Concern (VOC) including Alpha, Beta, Gamma, Delta, Omicron BA.1, Omicron BA.2 and Omicron BA.4/BA.5.
Catalog No. 50-227-2075