Learn More
Sino Biological SARS-CoV-2 NP-CoV MAb 20ul (Sino Biological)
Supplier:  Sino Biological 68097MM15020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Nucleocapsid CTD Protein (Catalog#40588-V07E5; YP_009724397.2; Lys248-Pro365). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1895