Learn More
Sino Biological SARS-CoV-2 NP-CoV PAb 100ul (Sino Biological)
Supplier:  Sino Biological 68098T30100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 Nucleocapsid NTD (Catalog#40588-V07E10; YP_009724397.2; Thr49-Gly175). SARS-CoV-2 Nucleocapsid NTD specific IgG was purified by SARS-CoV-2 Nucleocapsid NTD affinity chromatography.
Catalog No. 50-237-3150