Learn More
Cayman Chemical SARS-CoV-2 nsp1recombInt 500ug
SDP
Catalog No. 502944565
Encompass_Preferred
Source: Recombinant SARS-CoV-2 N-Terminal His-tagged nsp1 expressed in E. coli • Amino acids: 13-128
Catalog No. 50-294-4565 Supplier Cayman Chemical Supplier No. 40879500 UG
Source: Recombinant SARS-CoV-2 N-Terminal His-tagged nsp1 expressed in E. coli • Amino acids: 13-128