Learn More
Cayman Chemical SARS-CoV-2 nsp4recombInt 100ug
SDP
Catalog No. 502943169
Encompass_Preferred
Source: Recombinant SARS-CoV-2 N-terminal GST-tagged nsp4 expressed in E. coli • Amino acids: 405-500
Catalog No. 50-294-3169 Supplier Cayman Chemical Supplier No. 40880100 UG
Source: Recombinant SARS-CoV-2 N-terminal GST-tagged nsp4 expressed in E. coli • Amino acids: 405-500