Learn More
Medchemexpress LLC SARS-CoV-2 NSP7-NSP8 10ug | 10ug
SDP
Catalog No. 5000187550
Encompass_Preferred
The ORF1ab gene is a specifically expressed gene of SARS-CoV-2 and can be quickly and sensitively detected to indirectly indicate the level of SARS-CoV-2 The ORF1ab protein is associated with viral replication and transcription inhibition of host translation machinery and suppression of host immune responses SARS-CoV-2 NSP7-NSP8 Heterodimer Protein (His) is a recombinant protein dimer complex containing Virus-derived SARS-CoV-2 NSP7-NSP8 Heterodimer protein expressed by E coli with C-His labeled tag SARS-CoV-2 NSP7-NSP8 Heterodimer Protein (His) has molecular weight of 32 8 kDa
Catalog No. 50-001-87550 Supplier Medchemexpress LLC Supplier No. HYP7781610UG
The ORF1ab gene is a specifically expressed gene of SARS-CoV-2 and can be quickly and sensitively detected to indirectly indicate the level of SARS-CoV-2 The ORF1ab protein is associated with viral replication and transcription inhibition of host translation machinery and suppression of host immune responses SARS-CoV-2 NSP7-NSP8 Heterodimer Protein (His) is a recombinant protein dimer complex containing Virus-derived SARS-CoV-2 NSP7-NSP8 Heterodimer protein expressed by E coli with C-His labeled tag SARS-CoV-2 NSP7-NSP8 Heterodimer Protein (His) has molecular weight of 32 8 kDa