Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 NSP7 Recombinant Protein

Catalog No. PIRP88284
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88284 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88284 Supplier Invitrogen™ Supplier No. RP88284
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. Reconstituted with sterile deionized water to 0.25mg/mL Concentration is measured by UV.

Plays a role in viral RNA synthesis (PubMed:32358203, PubMed:32277040, PubMed:32438371, PubMed:32526208). Forms a hexadecamer with nsp8 (8 subunits of each) that may participate in viral replication by acting as a primase. Alternatively, may synthesize substantially longer products than oligonucleotide primers.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/mL
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation 20mM tris with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Molecular Weight (g/mol) 9.4 kDa
Name SARS-CoV-2 NSP7
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias COVID19 Nsp7; COVID-19 NSP7; SARS-CoV-2 Nonstructural Protein 7; SARS-CoV2 NSP7; SARS-COV-2 NSP7
Common Name SARS-CoV-2 NSP7
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence SARS-CoV-2 NSP7, amino acids Ser1-Gln83 (Accession # YP_009725303.1) with an N-terminal GP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.