Learn More
Cayman Chemical SARS-CoV-2 nsp7recombInt 500ug
SDP
Catalog No. 502945908
Encompass_Preferred
Source: Recombinant SARS-CoV-2 C-terminal His-tagged nsp7 expressed in E. coli • Amino acids: 3,831-3,913
Catalog No. 50-294-5908 Supplier Cayman Chemical Supplier No. 40881500 UG
Source: Recombinant SARS-CoV-2 C-terminal His-tagged nsp7 expressed in E. coli • Amino acids: 3,831-3,913