Learn More
Bon Opus Biosciences Recombinant SARS-CoV-2 NSP8 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP21550UG
Encompass_Preferred
Recombinant SARS-CoV-2 NSP8 is produced by our E.coli expression system and the target gene encoding Ala1-Gln198 is expressed with a 8His tag at the C-terminus. Expression system: E. coli. Accession# YP_009725304.1. Formulation: Supplied as a 0.2 um filtered solution of 20mM Tris, 150mM NaCl, 10%Glycerol, 4mM MgCl2, pH8.0. Bon Opus Cat# BP215
Catalog No. 50-208-0303