Learn More
Cayman Chemical SARS-CoV-2 nsp8recombInt 500ug
SDP
Catalog No. 502945102
Encompass_Preferred
Source: Recombinant SARS-CoV-2 N-terminal His- and DYKDDK-tagged nsp8 expressed in E. coli • Amino acids: 1-198
Catalog No. 50-294-5102 Supplier Cayman Chemical Supplier No. 40882500 UG
Source: Recombinant SARS-CoV-2 N-terminal His- and DYKDDK-tagged nsp8 expressed in E. coli • Amino acids: 1-198