Learn More
AdipoGen SARS-CoV-2 Nucleocapsid Protein (rec.) (His) (100 µg)
SDP
Supplier:  AdipoGen CHIB233501C100
Encompass_Preferred
Recombinant protein. SARS-CoV-2 Nucleocapsid protein (aa 1-419) is fused at the N-terminus to a His-tag. Source: E. coli. Endotoxin: <1EU/µg purified protein (LAL test).. Lyophilized. Contains PBS. SARS-CoV-2 shares 79.5% sequence identity with SARS-CoV and is 96.2% identical at the genome level to the bat coronavirus BatCoV RaTG133, suggesting it had originated in bats. The coronaviral genome encodes four major structural proteins: the Spike (S) protein, Nucleocapsid (N) protein, Membrane/Matrix (M) protein and the Envelope (E) protein. The SARS Envelope (E) protein has a role in viral budding and virion envelope morphogenesis. The SARS Membrane/Matrix (M) protein is one of the major structural viral proteins. It is an integral membrane protein involved in the budding of the viral particles and interacts with SARS Spike (S) protein and the Nucleocapsid (N) protein. The N protein contains two domains, both of them bind the virus RNA genome via different mechanisms.
Catalog No. 50-205-4695