Learn More
Sino Biological SARS-CoV-2 Nucleocapsid Antibody (Biotin), Rabbit PAb, Antigen Affinity Purified 20 µg
Catalog No. 5000750783
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 / 2019-nCoV Nucleocapsid Protein (Catalog#40588-V08B; YP_009724397.2(335Gly/Ala); Met1-Ala419) and then biotinylated. The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Nucleocapsid affinity chromatography and then biotinylated.
Catalog No. 50-007-50783 Supplier Sino Biological Supplier No. 40588T30B20
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 / 2019-nCoV Nucleocapsid Protein (Catalog#40588-V08B; YP_009724397.2(335Gly/Ala); Met1-Ala419) and then biotinylated. The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Nucleocapsid affinity chromatography and then biotinylated.