Learn More
Sino Biological SARS-CoV-2 Nucleocapsid Antibody, Mouse MAb 20 µL
Catalog No. 5000705165
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, SARS-CoV-2 Nucleocapsid (Catalog#40588-V08B; YP_009724397.2(335Gly/Ala); Met1-Ala419). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-05165 Supplier Sino Biological Supplier No. 40588MM15720
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, SARS-CoV-2 Nucleocapsid (Catalog#40588-V08B; YP_009724397.2(335Gly/Ala); Met1-Ala419). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.