Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Nucleocapsid CTD Protein (His Tag) 100ug
Supplier:  Sino Biological 40588V07E5100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid CTD (YP_009724397.2) (Lys248-Pro365) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-211-6500
 Restricted Item