Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (MALS verified)
Catalog No. 502107104
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (NUN-C5143) is expressed from E.coli cells. It contains AA Gly 44 - Glu 174 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (Cat. No. NUN-C5143) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (Cat. No. NUN-S61) with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-210-7104 Supplier ACROBiosystems Supplier No. NUNC51431MG
Recombinant Protein;1MG;SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (NUN-C5143) is expressed from E.coli cells. It contains AA Gly 44 - Glu 174 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 16 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Nucleocapsid Protein NTD, His Tag (Cat. No. NUN-C5143) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Mouse IgG1 (Cat. No. NUN-S61) with a linear range of 0.2-3 ng/mL (QC tested).