Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (MALS verified)
Catalog No. 502107084
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (NUN-C5145) is expressed from E.coli cells. It contains AA Ser 255 - Pro 364 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 13.4 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (Cat. No. NUN-C5145) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41A1) with a linear range of 0.1-1 ng/mL (QC tested).
Catalog No. 50-210-7084 Supplier ACROBiosystems Supplier No. NUNC5145100UG
Recombinant Protein;100UG;SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (NUN-C5145) is expressed from E.coli cells. It contains AA Ser 255 - Pro 364 (Accession # QHO62115.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 13.4 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).;Immobilized SARS-CoV-2 Nucleocapsid Protein CTD, His Tag (Cat. No. NUN-C5145) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41A1) with a linear range of 0.1-1 ng/mL (QC tested).