Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Nucleocapsid protein (D3L, R203K, G204R, S235F), His Tag
Catalog No. 502107375
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). D3L/ R203K/ G204R/ S235F were identified in the SARS-CoV-2 variant (known as 20B/501Y.V1, VOC 202012/01, or B.1.1.7 lineage) which emerged in the United Kindom.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.4 kDa. The protein migrates as 30-35 kDa and 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C52H8) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41A1) with a linear range of 0.06-1 ng/mL (QC tested).
Catalog No. 50-210-7375 Supplier ACROBiosystems Supplier No. NUNC52H8100UG
Recombinant Protein;100UG;SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). D3L/ R203K/ G204R/ S235F were identified in the SARS-CoV-2 variant (known as 20B/501Y.V1, VOC 202012/01, or B.1.1.7 lineage) which emerged in the United Kindom.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.4 kDa. The protein migrates as 30-35 kDa and 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 Nucleocapsid protein, His Tag (Cat. No. NUN-C52H8) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 (Cat. No. NUN-S41A1) with a linear range of 0.06-1 ng/mL (QC tested).