Learn More
STEMCELL Technologies SARS-CoV-2 (Nucleocapsid Protein) Peptide Pool, Size: 1 Unit
SDP
Supplier:  STEMCELL Technologies 1000647
Encompass
The SARS-CoV-2 (Nucleocapsid Protein) Peptide Pool is a lyophilized mixture of 102 peptides from the nucleocapsid protein of SARS-CoV-2. Nucleocapsid protein is one of four structural proteins that interacts with RNA to form the nucleocapsid. It also interacts with membrane protein (protein M) in the packaging of positive-strand viral genome RNA during virion assembly. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 - 419 on SARS-CoV-2 nucleocapsid protein.
Catalog No. 50-239-0138