Learn More
Genscript Corporation SARS-CoV-2 Nucleocapsid Protei
Supplier:  Genscript Corporation Z03732100
Encompass
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2) also known as 2019-nCoV is a positive-sense single-stranded RNA virus It caused coronavirus disease 2019 (COVID-19) Nucleocapsid Protein is the most abundant structural protein of the coronavirus which is associated with the nucleic acid The sublineage B 1 617 2 has been redesignated as a variant of concern (VOC-21APR-02) in May 2021 which spreads more quickly than the original version of the virus
Catalog No. 50-000-57188