Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ SARS-CoV-2 Nucleoprotein (Omicron,B.1.1.529) ELISA Kit
Shop All MP Biomedicals, Inc ProductsDescription
This ELISA kit is for in vitro quantitative determination of SARS-CoV-2 (Omicron, B.1.1.529) nucleoprotein concentrations in plasma, tissue homogenates, and other biological fluids from various mammalian species.
Specifications
Specifications
| Content And Storage | -20°C |
| Assay | EIA/ELISA |
| Immunoassay Kit Format | 96-well Plate |
| Measuring Range | Detection range: 0.156 to 10 ng/mL; Detection sensitivity: 0.094 ng/mL |
| Quantity | 96 Tests |
| Regulatory Status | RUO |
For research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?