Learn More
Cayman Chemical SARS-CoV-2 nutralIzng ntIb 1ea
SDP
Catalog No. 502895243
Encompass_Preferred
A set of SARS-CoV-2 neutralizing antibody human serum controls containing negative control serum, as well as positive control serum spiked with low, mid, and high levels of recombinant human anti-SARS-CoV-2 neutralizing IgG antibody
Catalog No. 50-289-5243 Supplier Cayman Chemical Supplier No. 5020901 EA
A set of SARS-CoV-2 neutralizing antibody human serum controls containing negative control serum, as well as positive control serum spiked with low, mid, and high levels of recombinant human anti-SARS-CoV-2 neutralizing IgG antibody