Learn More
Cayman Chemical SARS-CoV-2 nutralIzng ntIb 1UG
SDP
Catalog No. 502896039
Encompass_Preferred
SARS-CoV-2 Neutralizing Antibody Standard has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Neutralizing Antibody Detection ELISA Kit (Item No. 502070).
Catalog No. 50-289-6039 Supplier Cayman Chemical Supplier No. 5020741 UG
SARS-CoV-2 Neutralizing Antibody Standard has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Neutralizing Antibody Detection ELISA Kit (Item No. 502070).