Learn More
Cayman Chemical SARS-CoV-2 nutralIzng ntIb 1UG
SDP
Supplier:  Cayman Chemical 5022231 UG
Encompass_Preferred
SARS-CoV-2 Neutralizing Antibody Standard has been formulated and tested to work with Cayman’s Assay Kits. Detailed instructions for its use are contained in the respective Cayman Assay Kit booklet.
Catalog No. 50-289-5217