Learn More
Sino Biological SARS-CoV-2 Omicron (B.1.1.529) Nucleocapsid Antibody, Mouse MAb 20 µL
Catalog No. 5000786134
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, SARS-CoV-2 Omicron (B.1.1.529) Nucleocapsid (Catalog#40588-V07E34; YP_009724397.2; Met1-Ala419). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-007-86134 Supplier Sino Biological Supplier No. 68270MM28520
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, SARS-CoV-2 Omicron (B.1.1.529) Nucleocapsid (Catalog#40588-V07E34; YP_009724397.2; Met1-Ala419). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.