Learn More
ACROBiosystems SARS-CoV-2 ORF8 Protein, His Tag (OR8-C52H1) is expressed from human 293 cells (HEK293). It contains AA Phe 16 - Ile 121 (Accession # P0DTC8-1).
Supplier:  ACROBiosystems OR8C52H1100UG
Encompass_Preferred
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.3 kDa. The protein migrates as 18-21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-223-1038