Learn More
Medchemexpress LLC SARS-CoV-2 Plpro/papain-like protease protein (His) | >90.0% | 50 UG
SDP
Catalog No. 5000291158
Encompass_Preferred
Recombinant His-tagged SARS-CoV-2 papain-like protease (PLpro) supplied as a purified protein for biochemical and enzymatic studies. Supplied as 50 μg of purified protein expressed in E. coli and characterized by reducing SDS-PAGE; intended for enzymology, inhibitor screening, and binding assays.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • N-terminal His tag for affinity purification and detection.
  • Expressed in E. coli for scalable production.
  • Approximate molecular weight 36.8 kDa for identity confirmation.
  • Supplied frozen; store at -80°C and avoid repeated freeze-thaw cycles.
  • Suitable for enzymatic assays, inhibitor screening, and biochemical characterization.
Catalog No. 50-002-91158 Supplier Medchemexpress LLC Supplier No. HYP7602150UG
Recombinant His-tagged SARS-CoV-2 papain-like protease (PLpro) supplied as a purified protein for biochemical and enzymatic studies. Supplied as 50 μg of purified protein expressed in E. coli and characterized by reducing SDS-PAGE; intended for enzymology, inhibitor screening, and binding assays.
  • High purity (>90%) as determined by reducing SDS-PAGE.
  • N-terminal His tag for affinity purification and detection.
  • Expressed in E. coli for scalable production.
  • Approximate molecular weight 36.8 kDa for identity confirmation.
  • Supplied frozen; store at -80°C and avoid repeated freeze-thaw cycles.
  • Suitable for enzymatic assays, inhibitor screening, and biochemical characterization.