Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ SARS-CoV-2 Process Control
SDP

Catalog No. 23047375 Shop All Microbiologics Products
Change view
Click to view available options
Type:
Lyophilized Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-047-375 Lyophilized Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-047-375 Supplier Microbiologics™ Supplier No. R9000

missing translation for 'validMainframeMsg'

SARS-CoV-2 Process Control can be used for SARS-CoV-2 assay development, full process control (including extraction), sample adequacy control and spike-in controls.

  • SARS-CoV-2 Process Control is made with RNA transcripts containing diagnostically relevant targets (CDC and WHO consensus sequences) encapsulated in a phage protein.
  • Nucleotides contained in each pellet/swab (All positions in reference to GenBank Acc. NC_045512.2)
  • Orf1ab/RdRP: 12,690 ..12,797 | 13,342 ..13,460 | 14,080 ..14,186 | 15,431 ..15,530
  • S (Spike): 18,778 ..18,909 | 24,354 ..24,900
  • E (Envelope): 26,245 ..26,427 | 26,269 ..26,381
  • N (Nucleocapsid): 28,237 ..29,280
  • Includes human respiratory epithelial cells
  • RUO

Specifications

Biosafety Level 1
Type Lyophilized Pellets
Packaging Plastic Box; Individually packaged contents
Quantity 6/Pk.
Source United States
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.