Learn More
Sigma Aldrich Fine Chemicals Biosciences SARS CoV 2 Receptor Bindin
Catalog No. 502905251
Encompass
SARS-CoV-2 belongs to Coronaviridae (CoVs) - a large family of viruses that usually cause mild to moderate upper-respiratory tract diseases.1 CoVs are single-stranded RNA viruses. They have a high mutation rate and are therefore highly diverse.1 Their hosts include human and non-human mammals as well as birds.1
Catalog No. 50-290-5251 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE100050UG
SARS-CoV-2 belongs to Coronaviridae (CoVs) - a large family of viruses that usually cause mild to moderate upper-respiratory tract diseases.1 CoVs are single-stranded RNA viruses. They have a high mutation rate and are therefore highly diverse.1 Their hosts include human and non-human mammals as well as birds.1