Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 S protein (F817P, A892P, A899P, A942P, K986P, V987P), His Tag (MALS verified)
Supplier:  ACROBiosystems SPNC52HD500UG
Encompass_Preferred
Recombinant Protein;500UG;SARS-CoV-2 S protein, His Tag (SPN-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 138.2 kDa. The protein migrates as 75-85 kDa, 90-100 kDa and 150-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S protein, His Tag (Cat. No. SPN-C52Hd) at 1 μg/mL (100 μL/well) can bind Human ACE2, Fc Tag (Cat. No. AC2-H5257) with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-210-7372