Learn More
Medchemexpress LLC SARS-CoV-2 S protein receptor-binding domain, biotinylated (Omicron B.1.1.529) | 96.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7444610UG
Encompass_Preferred
SARS-CoV-2 S protein receptor-binding domain, biotinylated (Omicron B.1.1.529), expressed in HEK293 cells with C-terminal Avi and His tags. Supplied as a 10 μg aliquot for research use; the certificate of analysis reports 96% purity by reducing SDS-PAGE.
  • Biotinylated at Avi tag for site-specific immobilization.
  • Expressed in mammalian cells for native glycosylation.
  • C-terminal His tag for facile purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Suitable for ELISA, SPR, BLI, and antibody binding studies.
  • Supplied in a stable formulation for storage and handling.
Catalog No. 50-002-93871