Learn More
Medchemexpress LLC SARS-CoV-2 S trimer protein (XBB.1.16, HEK293, His) | >90.0% | 50UG
SDP
Catalog No. 5000174054
Encompass_Preferred
Recombinant SARS-CoV-2 spike (S) trimer corresponding to the XBB.1.16 variant, expressed in HEK293 cells and supplied as a C-terminal His-tagged soluble trimer in a buffered, filtered solution. Intended for research use in antigenicity, binding, and immunoassay applications; low endotoxin levels and recommended frozen storage preserve biological activity.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag facilitates affinity purification and detection.
  • Supplied as a 0.22 μm filtered, buffered solution ready for assays.
  • Endotoxin level <1 EU/μg suitable for sensitive bioassays.
  • High purity (>90%) as determined by Tris-Bis PAGE.
  • Stable when stored frozen; ship on dry ice and avoid repeated freeze-thaw cycles.
Catalog No. 50-001-74054 Supplier Medchemexpress LLC Supplier No. HYP70282150UG
Recombinant SARS-CoV-2 spike (S) trimer corresponding to the XBB.1.16 variant, expressed in HEK293 cells and supplied as a C-terminal His-tagged soluble trimer in a buffered, filtered solution. Intended for research use in antigenicity, binding, and immunoassay applications; low endotoxin levels and recommended frozen storage preserve biological activity.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag facilitates affinity purification and detection.
  • Supplied as a 0.22 μm filtered, buffered solution ready for assays.
  • Endotoxin level <1 EU/μg suitable for sensitive bioassays.
  • High purity (>90%) as determined by Tris-Bis PAGE.
  • Stable when stored frozen; ship on dry ice and avoid repeated freeze-thaw cycles.