Learn More
Bon Opus Biosciences Recombinant SARS-CoV-2 (2019-nCoV) S1 Protein (C-mFc) 500ug
SDP
Supplier:  Bon Opus Biosciences DRA30500UG
Encompass_Preferred
Recombinant SARS-CoV-2 (2019-nCoV) S1 Protein is produced by our Mammalian expression system and the target gene encoding Gln14-Arg685 is expressed with a mFc tag at the C-terminus. Accession# QHD43416.1. Formulation: Supplied as a 0.2 μM filtered solution of PBS, pH 7.4. Bon Opus Cat# DRA30
Catalog No. 50-217-2837