Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G), Fc Tag
Supplier:  ACROBiosystems S1DC5256100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1). The mutations (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G) were identified in the SARS-CoV-2 variant (known as B.1.351 or 20C/501Y.V2) which emerged in the South Africa.;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101.1 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, His Tag (Cat. No. AC2-H52H8) at 2 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein, Fc Tag (Cat. No. S1D-C5256) with a linear range of 2-25 ng/mL (QC tested).
Catalog No. 50-210-7206