Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 S1 protein NTD (L18F, T20N, P26S, D138Y, R190S), His Tag (MALS verified)
Supplier:  ACROBiosystems S1DC52HE1MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S1 protein NTD, His Tag (S1D-C52He) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1). The L18F/ T20N/ P26S/ D138Y/ R190S mutations were identified on the spike protein N-terminal domain (NTD) in the SARS-CoV-2 variant (known as P.1) which emerged in Brazil.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S1 protein NTD, His Tag (Cat. No. S1D-C52He) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike NTD Antibody, Chimeric mAb (Cat. No. SPD-M121) with a linear range of 0.1-0.8 ng/mL (QC tested).;Immobilized Anti-SARS-CoV-2 Spike NTD Antibody, Chimeric mAb (Cat. No. SPD-M121) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 S1 protein NTD, His Tag (Cat. No. S1D-C52He) with a linear range of 4-31 ng/mL (Routinely tested).
Catalog No. 50-210-7331