Learn More
AdipoGen SARS-CoV-2 S1 RBD His
SDP
Catalog No. 502562894
Encompass_Preferred
Recombinant protein. Receptor-binding domain RBD of SARS-CoV-2 Spike protein S1 aa 319-541 is fused at the C-terminus to a His-tag. Binds to anti-SARS-CoV-2 Spike RBD antibodies in serum or plasma. Endotoxin <0.01EU/µg purified protein LAL test. Lyophilized. Contains PBS. SARS-CoV-2 shares 79.5% sequence identity with SARS-CoV and is 96.2% identical at the genome level to the bat coronavirus BatCoV RaTG133, suggesting it had originated in bats. The coronaviral genome encodes four major structural proteins the Spike S protein, Nucleocapsid N protein, Membrane/Matrix M protein and the Envelope E protein. The SARS Envelope E protein contains a short palindromic transmembrane helical hairpin that seems to deform lipid bilayers, which may explain its role in viral budding and virion envelope morphogenesis. The SARS Membrane/Matrix M protein is one of the major structural viral proteins.
Catalog No. 50-256-2894 Supplier AdipoGen Supplier No. AG40B0195C050
Recombinant protein. Receptor-binding domain RBD of SARS-CoV-2 Spike protein S1 aa 319-541 is fused at the C-terminus to a His-tag. Binds to anti-SARS-CoV-2 Spike RBD antibodies in serum or plasma. Endotoxin <0.01EU/µg purified protein LAL test. Lyophilized. Contains PBS. SARS-CoV-2 shares 79.5% sequence identity with SARS-CoV and is 96.2% identical at the genome level to the bat coronavirus BatCoV RaTG133, suggesting it had originated in bats. The coronaviral genome encodes four major structural proteins the Spike S protein, Nucleocapsid N protein, Membrane/Matrix M protein and the Envelope E protein. The SARS Envelope E protein contains a short palindromic transmembrane helical hairpin that seems to deform lipid bilayers, which may explain its role in viral budding and virion envelope morphogenesis. The SARS Membrane/Matrix M protein is one of the major structural viral proteins.