Learn More
AdipoGen SARS-CoV-2 S1 RBD mAb AB68
SDP
Supplier:  AdipoGen AG27B6311C050
Encompass_Preferred
Recombinant Antibody. Recognizes the SARS-CoV-2 S1 Receptor-binding domain. Does not cross-react with HCoV-OC43, HCoV-229E, HCoV-NL63, HCoV-HKU1, MERS-CoV or SARS-CoV. Applications ELISA, ICC, WB. Clone AB68-A09. Isotype Mouse IgG2a. Formulation Liquid. In PBS. Coronaviruses CoVs are enveloped non-segmented positive-sense single-stranded RNA viruses and can infect respiratory, gastrointestinal, hepatic and central nervous system of human and many other wild animals. Recently, a new severe acute respiratory syndrome beta-coronavirus called SARS-CoV-2 or 2019-nCoV has emerged, which causes an epidemic of acute respiratory syndrome called coronavirus human disease 2019 or COVID-19. SARS-CoV-2 shares 79.5% sequence identity with SARS-CoV and is 96.2% identical at the genome level to the bat coronavirus BatCoV RaTG133, suggesting it had originated in bats.
Catalog No. 50-256-3132