Learn More
Medchemexpress LLC SARS-CoV-2 S2 HEK29 1mg | 1mg
SDP
Catalog No. 5000187105
Encompass_Preferred
The SARS-Cov-2 S glycoprotein is a SARS-Cov-2 S glycoprotein The amino acid sequence 1261-1267a a of the SARS-Cov-2 S glycoprotein is transmembrane The SARS-Cov-2 S glycoprotein is a highly glycosylated trimer each of which consists of 1260 amino acids (residues 14-1273) divided into four structural domains the n-terminal domain (NTD) the c-terminal domain (CTD also known as the receptor-binding domain RBD) and two subdomains (SD1 and SD2) SARS-CoV-2 S2 Protein (HEK293 His) is the recombinant Virus-derived SARS-CoV-2 S2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87105 Supplier Medchemexpress LLC Supplier No. HYP774811MG
The SARS-Cov-2 S glycoprotein is a SARS-Cov-2 S glycoprotein The amino acid sequence 1261-1267a a of the SARS-Cov-2 S glycoprotein is transmembrane The SARS-Cov-2 S glycoprotein is a highly glycosylated trimer each of which consists of 1260 amino acids (residues 14-1273) divided into four structural domains the n-terminal domain (NTD) the c-terminal domain (CTD also known as the receptor-binding domain RBD) and two subdomains (SD1 and SD2) SARS-CoV-2 S2 Protein (HEK293 His) is the recombinant Virus-derived SARS-CoV-2 S2 protein expressed by HEK293 with C-His labeled tag