Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 S2 protein (T716I, S982A, D1118H), His Tag
Supplier:  ACROBiosystems S2NC52HD1MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S2 protein (T716I, S982A, D1118H), His Tag (S2N-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P/ A892P/ A899P/ A942P/ K986P/ V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein. The T716I/ S982A/ D1118H mutations were identified in the SARS-CoV-2 variant (known as B.1.1.7, 20B/501Y.V1 or VOC 202012/01) which emerged in the United Kingdom.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.6 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 S2 protein (T716I, S982A, D1118H), His Tag (Cat. No. S2N-C52Hd) at 1 μg/mL (100 μL/well) can bind Anti-SARS-CoV-2 Spike S2 p
Catalog No. 50-210-7309