Learn More
Genscript Corporation SARS-CoV-2 Spike 484Q Antibo
Supplier:  Genscript Corporation A02155100
Encompass
SARS-CoV-2 Spike (484Q) Antibody (Q8) mAb Mouse is specific for 484Q site of B 1 617 SARS-CoV-2 This product can recognize single mutant (E484Q) and double mutant (L452R E484Q) but can not recognize the wild type and single mutant (L452R)
Catalog No. 50-000-58893