Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike Neutralizing Antibody, Mouse Mab
Supplier:  Sino Biological 40592MM57100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, Recombinant SARS-CoV-2 (2019-nCoV) Spike RBD-mFc Protein (Catalog#40592-V05H; YP_009724390.1; Arg319-Phe541). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. NC1823017