Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike Neutralizing Antibody, Mouse Mab
Supplier:  Sino Biological 40591MM43100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, Recombinant SARS-CoV-2 (2019-nCoV) Spike S1-mFc Protein (Catalog#40591-V05H1; YP_009724390.1; Val16-Arg685). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. NC1823016